This product is now discontinued, View Related Products
Flir Ax5-Series Fix Mount Thermal Cameras (only)
Get smooth, continuous automated processing
  • GenICam protocol supports seamless integration with camera
  • Built-in Gigabit Ethernet connection allows real-time 14-bit image streaming to computer
  • Easily synchronize between multiple cameras
The FLIR Ax5-Series thermal imaging cameras are ideal for continuous monitoring of temperatures in automation or machine vision environments. Operating in temperatures up to 140°F (60°C), these versatile, non-contact cameras are sensitive enough to detect temperature differences as small as 50 mK. Record crisp 640 x 512 pixel thermal images up to 30 Hz with the A65 model or stream 320 x 256 pixel thermal images up to 60 Hz with the A35 model.
These affordable cameras integrate seamlessly into your existing system for instant data analysis and provide temperature linear output through GenlCam™ compliant software. The series offers 10 field-of-view options, supports synchronization between cameras for stereoscopic applications, and features GigE Vision interface for fast image transfer using low-cost standard cables over long distances. Its compact size makes it ideal for installing in electrical cabinets and other small spaces.
$7,359.58 - $15,102.25CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Camera Included Detector Size Max Temperature (° C)
Availability Pricing
Flir A35 Automation Thermal Camera, FOV 24°, 19 mm Lens RK-39757-14 Mfr # 83213-0102
COMPARE
19 mm lens 320 x 356 550
$8,352.65CAD / Each
Flir A35 Automation Thermal Camera, FOV 63°, 7.5 mm Lens RK-39757-15 Mfr # A35 FOV 63
COMPARE
7.5 mm lens 320 x 356 550
$7,359.58CAD / Each
Flir A35 Automation Thermal Camera, FOV 48°, 9 mm Lens RK-39757-16 Mfr # A35 FOV 48
COMPARE
9 mm lens 320 x 356 550
$7,359.58CAD / Each
Flir A35 Automation Thermal Camera FOV 13°, 35 mm Lens RK-39757-17 Mfr # A35 FOV 13
COMPARE
35 mm lens 320 x 356 550
$7,801.60CAD / Each
Flir A35 Automation Thermal Camera, FOV 8°, 60 mm Lens RK-39757-18 Mfr # A35 FOV 8
COMPARE
60 mm lens 320 x 356 550
$7,801.60CAD / Each
Flir A65 Automation Thermal Camera, FOV 45°, 13 mm Lens RK-39757-19 Mfr # A65 FOV 45
COMPARE
13 mm lens 640 x 512 135
$12,684.41CAD / Each
Flir A65 Automation Thermal Camera, FOV 25°, 25 mm Lens RK-39757-20 Mfr # A65 FOV 25
COMPARE
25 mm lens 640 x 512 135
$12,684.41CAD / Each
Flir A65 Automation Thermal Camera, FOV 90°, 7.5 mm Lens RK-39757-21 Mfr # A65 FOV 90
COMPARE
7.5 mm lens 640 x 512 135
$12,684.41CAD / Each
Flir A65 Automation Thermal Camera, FOV 12°, 50 mm Lens RK-39757-22 Mfr # A65 FOV 12
COMPARE
50 mm lens 640 x 512 135
$11,779.75CAD / Each
Flir A65 Automation Thermal Camera, FOV 6°, 100 mm Lens RK-39757-23 Mfr # A65 FOV 6
COMPARE
100 mm lens 640 x 512 135
$15,102.25CAD / Each
Showing 1- 10 of 10