This product is now discontinued, View Related Products
Flir A310 PT Dual-Sensor Pan/Tilt Thermal Cameras
Detect potential threats before they happen
  • IP-66 rated housing
  • Day/night 36 x zoom color CCD
The A310 PT dual-sensor cameras combine thermal and visible light video security imaging with automated safety monitoring based on the detection of temperature measurement thresholds. By measuring slight changes in surface temperature, the cameras can detect hotspots or overheating equipment. An alarm alerts you if potential threats exceed pre-set levels.
Due to its composite video output, it is also an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
$36,827.38 - $44,194.33CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Detector Size Field Of View (FOV) Max Temperature (° F)
Availability Pricing
Flir 61002-1103 Pan and Tilt Automation Thermal Camera with 25 Deg Lens RK-39755-58 Mfr # 61002-1103
COMPARE
662
$36,827.38CAD / Each
Flir 61002-1104 Pan and Tilt Automation Thermal Camera with 45 Deg Lens RK-39755-59 Mfr # 61002-1104
COMPARE
662
$42,720.94CAD / Each
Flir A310 PT Pan and Tilt Automation Thermal Camera with 90° Lens RK-39755-61 Mfr # FLIR A310PT W/90 LENS
COMPARE
320 x 240 90° x 73° 662
$44,194.33CAD / Each
Flir A310 PT Pan and Tilt Automation Thermal Camera with 15° Lens RK-39755-62 Mfr # FLIR A310PT W/15 LENS
COMPARE
320 x 240 15° x 11.25° 662
$42,720.94CAD / Each
Flir A310 PT Pan and Tilt Automation Thermal Camera with 6° Lens RK-39755-63 Mfr # FLIR A310PT W/6 LENS
COMPARE
340 x 240 6° x 4.5° 662
$44,194.33CAD / Each