This product is now discontinued, View Related Products
Flir A3xxex Series Explosion Proof Thermal Cameras with IP67 Enclosure
Safely monitor critical assets in explosive atmospheres
  • Ideal for use in harsh, dusty environment-- IP67 rated
  • Heater prevents fogging and freezing of protection window
  • Plug and play installation
FLIR A310 ex is an ATEX compliant solution, with a thermal imaging camera mounted in an enclosure, making it possible to monitor critical and other valuable assets also in explosive atmospheres. Process monitoring, quality control and fire detection in potentially explosive locations are typical applications for the A310 ex. Explosive atmospheres need to be protected from ignition sources by selecting equipment and protective systems which meet the requirements of the ATEX Product Regulations or similar regulations. Flame-proof enclosure prevents any explosion transmission from the inside of the enclosure to the outside.
The integrated controller features several digital I/O channels and sensors for temperature, humidity and pressure. Among other functions, the I/O channels enable the user to switch on/off the camera and the heater via remote control. The access is accomplished through an integrated web interface or Modbus TCP/IP. The controller is equipped with two fiber optic and two Ethernet parts. This enables a flexible network integration in star or ring topologies.
Automation thermal imagers in many cases require application support prior to ordering. Please contact our technical application group or sales team for support on this product. In some areas of the country onsite field assessments can be arranged.
$29,460.43 - $33,291.25CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Field Of View (FOV) Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Flir 71001-1103 Explosion Proof Automation Thermal Camera, F=18mm and 25 Deg Lens RK-39755-31 Mfr # 71001-1103
COMPARE
25 degrees 662 320 x 240
$29,460.43CAD / Each
Flir 71001-1104 Explosion Proof Automation Thermal Camera, F=10mm and 45 Deg Lens RK-39755-32 Mfr # 71001-1104
COMPARE
45 degrees 662 320 x 240
$33,291.25CAD / Each