This product is now discontinued, View Related Products
Flir A3xx Series Fix Mount Thermal Cameras (only)
Offers accurate temperature measurement solutions at an affordable price
  • 320 x 240 pixel IR resolution
  • MPEG-4 streamed video
  • C-Video
These fixed-mounted thermal imagers can be installed almost anywhere to monitor your critical equipment. They offers an affordable and accurate temperature measurement solution for anyone who needs to solve problems that do not call for the highest speed or reaction and who uses a PC. Due to to its composite video output, it is also an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
Automation thermal imagers in many cases require application support prior to ordering. Please contact our technical application group or sales team for support on this product. In some areas of the country onsite field assessments can be arranged.
$12,610.75 - $16,575.64CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
FLIR A300 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens RK-39755-27 Mfr # 48201-1001
COMPARE
350 -20 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens
$12,610.75CAD / Each
FLIR A310 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens RK-39755-28 Mfr # 48201-1101
COMPARE
350 -20 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens with Built-in analysis function
$15,012.37CAD / Each
FLIR A315 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens RK-39755-29 Mfr # 48001-1101
COMPARE
350 -20 Automation Thermal Camera (320 x 240), F=18mm and 25 Deg Lens, GigE Compliant
$16,575.64CAD / Each