Idex® Flangeless Ferrules, 1/4-28 Flat-Bottom Connection for 1/16" OD Tubing
No need to flange tubing
  • Excellent for connecting rigid or semi-rigid tubing
Use with male nuts for a leak-tight connection.
$2.80 - $32.86CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Ferrule, Blue ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK RK-01939-30 Mfr # P-200X
COMPARE
Blue ETFE Flangeless Ferrule, Blue ETFE, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$28.58CAD / Pkg of 10
Idex Flangeless Ferrule, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK RK-02007-54 Mfr # P-200NX
COMPARE
Natural ETFE Flangeless Ferrule, 1/16\", 10/Pk
$32.86CAD / Pkg of 10
Idex Flangeless Ferrule, Blue ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA RK-02019-33 Mfr # P-200
COMPARE
Blue ETFE Flangeless Ferrule, Blue ETFE, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$3.32CAD / Each
Idex Flangeless Ferrule, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA RK-02019-34 Mfr # P-200N
COMPARE
Natural ETFE Flangeless Ferrule, Natural ETFE, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$3.24CAD / Each
Idex Flangeless Ferrule, Red ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA RK-02019-35 Mfr # P-200R
COMPARE
Red ETFE Flangeless Ferrule, Red ETFE, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$3.24CAD / Each
Idex Flangeless Ferrule, Natural PP, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA RK-02019-63 Mfr # P-221
COMPARE
Natural PP Flangeless Ferrule, Natural PP, 1/16\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.80CAD / Each