This product is now discontinued, View Related Products
Fixed Wavelength Detectors 22525
Replace your failed component or augment your liquid chromatography system with these precision UV/Vis detectors. All wetted parts are PEEK, PTFE, or Tefzel™ for excellent chemical resistance and biocompatibility. Integrate detector into your data stream via analog output.
Fixed Wavelength Detectors - use these durable detectors for repetitive analysis and preparative techniques. Standard 254 nm detection wavelength can be changed using optional filter conversion kits. Adapt for preparative operation using shorter path length flowcells (sold separately).
$3,489.72 - $3,679.05CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Max Wavelength (nm) Description
Availability Pricing
COMPARE
200 254 Fixed-Wavelength UV/VIS Detector, 254 nm; 115 VAC
$3,489.72CAD / Each
COMPARE
200 254 Fixed-wavelength detector, 254 nm. 230v (absorbance, wavelength)
$3,679.05CAD / Each