This product is now discontinued, View Related Products
Fischer Technology FMP Coating Thickness Gauge
Portable instruments deliver nondestructive, high precision measurements
  • Automatic probe recognition with USB port for data transfer to a PC
  • Units of measurement can be switched between µm and mils
  • Memory for up to 20,000 readings with Audible signal at measurement acquisition
  • Units display mean value, standard deviation, min, max, and range graphically. Probe not included and must be purchased seperately.
These easy-to-operate, rugged instruments are suitable for virtually any coating thickness measurement requirement. With a wide assortment of plug-in type probes, these meters are designed for nondestructive coating thickness measurements according to the magnetic induction method. In addition to adding up to 100 user-created calibration applications, the spectrum of requirements that this series is capable of spans from extensive painted specimens, to other very specific and complex measuring challenges. Select appropriate probes based on your application requirements. Probe not included and must be purchased seperately.
$2,378.05 - $4,128.44CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Measuring Method Width (in) Description
Availability Pricing
Fischer Technology FMP 30 Deltascope Coating Thickness Gauge RK-54100-03 Mfr # 605-022
COMPARE
Magnetic induction 7 Coating Thickness Gauge
$2,669.78CAD / Each
Fischer Technology FMP 30 Isoscope Coating Thickness Gauge RK-54100-04 Mfr # 605-028
COMPARE
Eddy current 7 Coating Thickness Gauge
$2,378.05CAD / Each
Fischer Technology FMP 40 Dualscope Coating Thickness Gauge RK-54100-12 Mfr # 605-024
COMPARE
Eddy current, Magnetic induction 7 Coating Thickness Gauge
$4,128.44CAD / Each