Sherwood Scientific Filter Top Caps for 5-L Sealed Tubs
Select cap 02658-68 for wetter or denser samples requiring more air flow. Use filter top cap 02658-69 for drying 5-µm particles.
Note: Only 100 g of sample can be dried at a time with filter top cap 02658-69.
$1,473.39 - $1,650.20CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sherwood Scientific 50035021 Filter top cap for 5 L sealed tubs, 250 mesh RK-02658-66 Mfr # 50035021
COMPARE
Filter top cap for 5 L sealed tubs, 250 mesh
$1,473.39CAD / Each
Sherwood Scientific 50035022 Filter top cap for 5-L sealed tub, 500 mesh RK-02658-67 Mfr # 50035022
COMPARE
Filter top cap for 5-L sealed tub, 500 mesh
$1,650.20CAD / Each