Saint Gobain® FEP Sampling Bags
Reusable bags resist damage from radiation and corrosive gases
  • Extra rugged—5 mil thick
  • Resists temperatures up to 200°F (93°C)
Bags feature an on/off valve with 3⁄16" nickel-plated brass hose barb or a a silicone septum in a polypropylene housing. Maxiumum temperature is 200°F (93°C).
$82.07 - $362.09CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Thickness (mil) Valve Volume (Liters)
Availability Pricing
Saint Gobain Gas Sampling Bag, FEP, 6" X 6", On/off Valve RK-01430-10 Mfr # D1075002
COMPARE
5 On/off 0.5
$103.73CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 6" X 10", On/off Valve RK-01430-20 Mfr # D1075001
COMPARE
5 On/off 1.2
$123.76CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 12" X 12", On/off Valve RK-01430-30 Mfr # D1075003
COMPARE
5 On/off 4.7
$123.76CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 24" X 36", On/off Valve RK-01430-40 Mfr # D1075004
COMPARE
5 On/off 50
$312.73CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 6" X 6", Septum Valve RK-01430-50 Mfr # D1075005
COMPARE
5 Septum 0.5
$82.07CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 6" X 10", Septum Valve RK-01430-60 Mfr # D1075006
COMPARE
5 Septum 1.2
$113.01CAD / Each
Saint Gobain Gas Sampling Bag, FEP, 24" X 36", Septum Valve RK-01430-80 Mfr # D1075008
COMPARE
5 Septum 50
$362.09CAD / Each
Showing 1- 7 of 7