This product is now discontinued, View Related Products

Extech Single Output Switching DC Power Supply

Excellent for bench technicians, engineers, and educational facilities
  • Three user-defined voltage and current presets provide additional flexibility
  • Dual-action rotary encoder control with push knob for easy fine and coarse tuning
  • Limit maximum current to 5 A through front panel auxiliary output and 10 A from the back panel
  • Remote control for output voltage/current and on/off
$707.21 - $777.94CAD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
Extech DCP60 DC Switching Power Supply, 600 W RK-20046-46 Mfr # DCP60
COMPARE
10 A 60 VDC ≤ 5 mVrms, < 100 mVp-p
$777.94CAD / Each
Extech DCP60-220 DC Switching Power Supply, 600 W, 220 VAC RK-20046-47 Mfr # DCP60-220
COMPARE
10 A 60 VDC ≤ 5 mVrms, < 100 mVp-p
$707.21CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat