Extech Pressure Meter/Datalogger Transducers
Get fast, realiable measurements
  • Ideal for air or non-corrosive/non-ionized gas
These interchangeable pressure transducers work well with the Extech SDL700 pressure meter/datalogger (model 68602-28).
$508.30CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Output Wetted Materials
Availability Pricing
Extech PT30 Pressure Transmitter, 30 psi RK-68602-30 Mfr # PT30
COMPARE
30 0 to 100mV DC Corrosion-resistant brass alloy
$508.30CAD / Each
Extech PT150 Pressure Transmitter, 150 psi RK-68602-32 Mfr # PT150
COMPARE
0 to 100mV DC Corrosion-resistant brass alloy
$508.30CAD / Each