Extech Compact temperature and humidity dataloggers
Place in small areas without AC outlets: refrigerators, freezers, vans, trucks, and storage crates
  • Records up to 16,000 readings, 8,000 for each parameter
This versatile, low-cost datalogger includes a real time clock so you can program an immediate or delayed start time. You can also program high and low set points. The LED lights on the front panel flash when the datalogger is recording and reaches the set points. Set the flexible datalogging interval to collect sample data from 2 to 2 hours at 1 second increments.
$324.13CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Humidity RH (RH) Max Temperature (° F) Temperature Accuracy (° F)
Availability Pricing
Extech 42275 Compact temperature and humidity datalogger kit RK-26842-90 Mfr # 42275
COMPARE
100 123 ±1 (-4 to 122F); ±2F (-40 to -4F, 123 to 185F)
$324.13CAD / Each