Extech 800 Series Digital Clamp-on Multimeters with built-in IR Thermometer
Clamp meter, multimeter and infrared thermometer all in one!
  • Locate hot spots and overheating equipment with built-in IR thermometer
Theses clamp-on multimeters priovide a built-in IR thermometer with laser pointer that makes troubleshooting heat sensitive areas simple. Features include data hold, Min/Max, autoranging with manual range button, auto power off, and an easy-to-read 4000 count backlit dual LCD display. Peak hold captures inrush currents and transients, and models 26834-22, -24 include true RMS and a type K thermometer probe. Units ar CAT III, 600V rated.
Model 26834-26 is CAT IV 600V rated.
$353.60CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy Description
Availability Pricing
Extech EX830 1000A AC/DC True RMS Clamp/DMM/IR Thermometer RK-26834-24 Mfr # EX830
COMPARE
600 ±1.5% + 2 digits 1000A AC/DC True RMS Clamp/DMM/IR Thermometer
$353.60CAD / Each