This product is now discontinued, View Related Products
Eutech Instruments pH 550 1/4-DIN Monitor
Small size, economical monitor with display
  • Measure and display both pH/ORP and temperature
  • Automatic or manual temperature compensation
  • Asymetrical or symmetrical mode operation
The pH 550 monitors are used to measure pH /ORP and temperature values in applications such as water treatment and monitoring, chemical processing, or food processing. The monitors have user-friendly features like menu-driven programs and calibrations that are easily performed with the push-button keypad. All setup and calibration parameters are retained in the nonvolatile memory to prevent data loss during power outages.
Measurements are compensated for temperature variations automatically when used with pH electrodes that have temperature elements. The monitors also offer manual temperature compensation with independent settings for calibration and process temperature. One- or two-point calibrations can be performed with USA or NIST buffer sets. All values, operational, and error messages are indicated on the large, dual-line LCD. All setup and calibration parameters are retained in the nonvolatile memory to prevent data loss during power outages.
$804.47 - $948.86CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH 550 pH/orp Monitor
$804.47CAD / Each
Eutech Instruments pH 550 pH/ORP Monitor with NIST-Traceable Calibration RK-56717-37 Mfr # 5671730NIST
COMPARE
pH 550 pH/ORP Monitor with NIST-traceable Calibration
$948.86CAD / Each