This product is now discontinued, View Related Products
Eurotherm Digital Temperature 2200E-Series Controllers
General Purpose PID Temperature Controllers
The 2200E series are configurable for PID, On/ Off or motorized valve control for heat only or heat/cool applications, satisfying both electrical and gas heating requirements. Self-tuning and customized cooling algorithms are included to optimize control performance. These units employ unique INSTANT ACCURACY™ technology on scaleable universal inputs accepting nine standard thermocouple and RTD inputs, DC process inputs or downloadable custom inputs. A simple ramp and hold profile can be programmed using the internal ramp generator and dwell timer. With custom cooling algorithms, these controllers are ideal for plastics extrusion.
$184.17 - $948.86CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Eurotherm 2204ECCVHH7THDHDLXX2XXENG controller configuration 1/4 Controller, DC, Rly, Tri RK-09081-11 Mfr # 2204ECCVHH7THDHDLXX2XXENG
COMPARE
1/4
$948.86CAD / Each
Eurotherm 2204ECCVHT1T1RFXX2XXENG controller configuration 1/4 Controller, Tri, Rly RK-09081-14 Mfr # 2204ECCVHT1T1RFXX2XXENG
COMPARE
1/4
$875.19CAD / Each
Eurotherm 2204E/CC/VH/R1/R1/XX/ controller configuration 1/4 Controller, Rly RK-09081-17 Mfr # 2204E/CC/VH/R1/R1/XX/
COMPARE
1/4
$829.52CAD / Each
Eurotherm 2216ECCVHLHXXFH2XXENG controller configuration 1/16 Controller, Log, Rly RK-09081-53 Mfr # 2216ECCVHLHXXFH2XXENG
COMPARE
1/16
$561.36CAD / Each
Eurotherm 2216ECCVHRHRCXX2XXENG controller configuration 1/16 Controller, Rly RK-09081-60 Mfr # 2216ECCVHRHRCXX2XXENG
COMPARE
1/16
$584.94CAD / Each
Eurotherm 2216ECCVHTHRCXX2XXENG controller configuration 1/16 Controller, Tri, Rly RK-09081-64 Mfr # 2216ECCVHTHRCXX2XXENG
COMPARE
1/16
$584.94CAD / Each
Eurotherm 2216ECCVHRHRCALXX2XX/ENG controller configuration 1/16 Controller RK-09082-08 Mfr # 2216ECCVHRHRCALXX2XX/ENG
COMPARE
1/16
$630.61CAD / Each
Eurotherm 2208ECCVHLHXXFHXXXX2FEENG controller configuration 1/8 Controller RK-09082-09 Mfr # 2208ECCVHLHXXFHXXXX2FEENG
COMPARE
1/8
$873.72CAD / Each
Eurotherm 2208E Temperature Controller, high & low alarm relay outputs RK-09083-05 Mfr # 2208E/CC/VH/LH/XX/FH/FL/XX/2XX.ENG
COMPARE
1/8
$773.53CAD / Each
Eurotherm 2208E Temperature Controller, cooling relay output RK-09083-18 Mfr # 2208E/CC/VH/LH/RC/RF/RF/XX/2XX/ENG
COMPARE
1/8
$813.31CAD / Each
Eurotherm Temperature Controller, heating/cooling relays RK-90440-47 Mfr # 2216E/CC/VH/RH/RC/XX/XX/2XX/ENG
COMPARE
1/16
$623.24CAD / Each
Eurotherm COMMS BOARDEI-BISY RS422 RK-94470-29 Mfr # SUB22/2FE
COMPARE
$184.17CAD / Each
Eurotherm COMMS BOARDMBUS RTU EIA485 RK-94470-31 Mfr # SUB22/2YM
COMPARE
$184.17CAD / Each
Showing 1- 13 of 13