BYK-Gardner Ergonomic Color Meter Kits
Precise measurements in a single, portable device
  • Color and 60° gloss measurement in one
  • Simple data transfer to Excel
  • Innovative LED technology
Use the color kits to measure the exact colors of plastics, textiles, foodstuffs, and other products for quality assurance. Ideal for quick pass/fail decisions in production.
$21,400.99 - $23,603.71CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Range Description
Availability Pricing
BYK-Gardner CC-6801 Ergonomic Color/Gloss Meter; 45/0 Color Geometry, 60 Gloss Geometry RK-59720-02 Mfr # CC-6801
COMPARE
LCD Color geometry: 45/0; Gloss geometry: 60° Ergonomic Color and Gloss Meter; 45°/0° Color Geometry, 60° Gloss Geometry
$21,400.99CAD / Each
BYK-Gardner CC-6807 Ergonomic Color Meter Kit, 45°/0° geometry RK-59720-05 Mfr # CC-6807
COMPARE
LCD Color geometry: 45/0 Ergonomic Color Meter Kit; 45°/0° Color Geometry
$23,603.71CAD / Each