Eppendorf Multi-Purpose 5804 Non-Refrigerated Centrifuges
Versatile rotor and adapter options can be used for virtually any application!
  • A variety of rotors and adapters for flexibility in the lab
Use these multi-purpose centrifuges for a variety of applications: high- speed separation of cell lysates. Rotor sensing feature automatically sets safe maximum speed. Order rotors and adapters separately.
Model 5804 with medium capacity can accommodate up to 4 x 100 mL bottles. This model also accepts swing-bucket rotors for deepwell plates for increased versatility.
$8,262.77CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Eppendorf 022622501 Multi-purpose centrifuge; 4 x 100 mL capacity, 115 VAC RK-02570-10 Mfr # 022622501
COMPARE
115
$8,262.77CAD / Each
Eppendorf 022622552 Multi-purpose centrifuge; 4 x 100 mL capacity, 220 VAC 02570-15 RK-02570-15 Mfr # 022622552
COMPARE
220
$8,262.77CAD / Each
Eppendorf 22 62 250-1 Multi-purpose centrifuge; 4 x 100 mL capacity, 115 VAC RK-02570-30 Mfr # 22 62 250-1
COMPARE
115
$8,262.77CAD / Each