Eppendorf Biopur tubes
Eppendorf® Biopur™ tubes have all the features of the standard and Safe-Lock™ tubes and are individually wrapped and certified sterile. Biopur™ tubes are pyrogen-, RNase-, DNA-and ATP-free; supplied with a quality assurance card.
$53.04 - $88.40CAD / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Color
Availability Pricing
Eppendorf 22-60-000-1 Microcentrifuge Tube, 0.5 mL, Natural Color RK-02560-32 Mfr # 22-60-000-1
COMPARE
0.5 Natural
$53.04CAD / Pkg of 50
Eppendorf 22-60-002-8 Microcentrifuge Tube, 1.5 mL, Natural Color RK-02560-35 Mfr # 22-60-002-8
COMPARE
1.5 Natural
$76.62CAD / Pkg of 100
Eppendorf 22-60-004-4 Microcentrifuge Tube, 2.0 mL, Natural Color RK-02560-38 Mfr # 22-60-004-4
COMPARE
2 Natural
$88.40CAD / Pkg of 100