Eppendorf 5810 Series Multi-Purpose Non-Refrigerated Centrifuges
Versatile rotor and adapter options can be used for virtually any application!
  • Superior quality and metal safety containment construction with aerosol tight lid options ensures your safety
  • Software control guarantees high accuracy, speed and temperature for optimal performance
Use these high-speed, multi-purpose centrifuges for a variety of applications: cell harvesting, high-speed separation of cell lysates, and DNA precipitations. The automatic rotor sensing feature will recognize your rotors and detect imbalances for safe operation. Order rotors and adapters separately.
Model 5810 accept up to 4 x 500 mL bottles, 28 x 50 mL conical tubes, or 16 x microplates — ideal for high throughput labs needing flexibility in sample running.
$10,487.59CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Eppendorf 022625004 Multi-purpose centrifuge 5810; 4 x 400 mL capacity, 120 VAC RK-02570-01 Mfr # 022625004
COMPARE
115
$10,487.59CAD / Each
Eppendorf 022625055 Multi-purpose centrifuge 5810; 4 x 400 mL capacity, 230 VAC RK-02570-02 Mfr # 022625055
COMPARE
230
$10,487.59CAD / Each