Eppendorf 5810 Series Multi-Purpose Centrifuge Tube Adapters
Choose the right centrifuge tube adapter for your application needs.
$475.90CAD / Pkg of 2
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Adapter Shape Type Tube Capacity
Availability Pricing
Eppendorf 022638700 Adapter for Swing Bucket (4 x 500 mL) Rotor 02570-67, 1.8-7 mL Tubes RK-02570-43 Mfr # 022638700
COMPARE
Flat with rubber mat 25 x 2.6 to 7 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638721 Adapter for Swing Bucket (4 x 500 mL) Rotor 02570-67, 4-10 mL Tubes RK-02570-47 Mfr # 022638721
COMPARE
Flat with rubber mat 16 x 4 to 10 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638742 Adapter for Swing Bucket (4 x 500 mL) Rotor 02570-67, 15 mL Conical Tubes RK-02570-96 Mfr # 022638742
COMPARE
Conical 12 x 15 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638769 Adapter for Swing Bucket (4 x 500 mL) Rotor 02570-67, 50 mL Conical Tubes RK-02570-97 Mfr # 022638769
COMPARE
Conical 5 x 50 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638386 Centrifuge Rotor Adapter (4 x 500 mL) Rotor 02570-69, 30-50 mL Flat bottom RK-02570-98 Mfr # 022638386
COMPARE
Flat with rubber mat 4 x 30 to 50 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638343 Adapter for Swing Bucket (4 x 500 mL) Rotor 02570-69, 50 mL Conical Tubes RK-02570-99 Mfr # 022638343
COMPARE
Conical 3 x 50 mL tubes per adapter
$475.90CAD / Pkg of 2
Eppendorf 022638343 Centrifuge Rotor 02570-69 Adapter, 1.8 to 7 mL Conical Tubes; 2/Pk RK-02571-49 Mfr # 022638246
COMPARE
Flat with rubber mat 15 x 1.8 to 7 mL tubes per adapter
$475.90CAD / Each
Eppendorf 022638343 Centrifuge Rotor 02570-69 Adapter, 4 to 10 mL Conical Tubes; 2/Pk RK-02571-51 Mfr # 022638262
COMPARE
Flat with rubber mat 12 x 4 to 10 mL tubes per adapter
$475.90CAD / Each
Eppendorf 022638343 Centrifuge Rotor 02570-69 Adapter, 15 mL Conical Tubes; 2/Pk RK-02571-53 Mfr # 022638289
COMPARE
Conical 9 x 15 mL tubes per adapter
$475.90CAD / Each
Showing 1- 9 of 9