Eppendorf 5804R Refrigerated Multi-Purpose Centrifuges
  • A variety of rotors and adapters for flexibility in the lab
Use these multi-purpose centrifuges for a variety of applications: cell harvesting, high- speed separation of cell lysates, and DNA precipitations. Rotor sensing feature automatically sets safe maximum speed. Refrigerator units cool rotor chamber in about 15 minutes and keep chambers at set temperature while lid is down. Order rotors and adapters separately.
Model 5804R with medium capacity can accommodate up to 4 x 100 mL bottles. This model also accepts swing-bucket rotors for deepwell plates for increased versatility.
$13,873.44CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Eppendorf 022623508 Refrigerated multi-purpose centrifuge; 4 x 250 mL capacity, 115 VAC RK-02570-20 Mfr # 022623508
COMPARE
115
$13,873.44CAD / Each
Eppendorf 022623559 Refrigerated multi-purpose centrifuge; 4 x 250 mL capacity, 220 VAC RK-02570-25 Mfr # 022623559
COMPARE
220
$13,873.44CAD / Each
Eppendorf 22 62 350-8 Refrigerated multi-purpose centrifuge; 4 x 100 mL capacity, 115 VAC RK-02570-40 Mfr # 22 62 350-8
COMPARE
115
$13,873.44CAD / Each
Eppendorf 22 62 355-9 Refrigerated multi-purpose centrifuge; 4 x 100 mL capacity, 220 VAC RK-02570-45 Mfr # 22 62 355-9
COMPARE
220
$13,873.44CAD / Each