Eppendorf 5424/5425R Variable Speed Microcentrifuge Rotors
Optional rotors add versitility
These optional rotors are compatible with the Eppendorf 5424/5424R variable-speed microcentrifuges. Centrifuge sold separately.
Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise.
$1,172.82 - $2,101.05CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Tubes Accepted Rotor Type Tube Sizes Accepted
Availability Pricing
Eppendorf 022653083 Spin Column Rotor Kit 18 x 1.5/2.0 mL, including lid RK-02580-50 Mfr # 022653083
COMPARE
18 Fixed-angle 1.5 or 2.0 mL microtubes
$1,172.82CAD / Each
Eppendorf 022653121 Fixed-angle 4 x 8-tube PCR-strips Rotor, with lid RK-02580-52 Mfr # 022653121
COMPARE
4 Fixed-angle 8-tube PCR strips
$1,172.82CAD / Each
Eppendorf 022653041 Fixed-angle 24 x 1.5/2.0 mL Rotor with chemical resistant (PTFE) coating RK-02580-54 Mfr # 022653041
COMPARE
24 Fixed-angle 1.5 or 2.0 mL microtubes
$1,670.82CAD / Each
Eppendorf 022636146 6 x PCR-strip rotor (30 x 1.5 to 2.0 mL tubes) RK-17483-53 Mfr # 022636146
COMPARE
30 Fixed-angle 1.5 or 2.0 mL microtubes
$2,101.05CAD / Each