Environmental Express/ Oakton® High-Performance Combination pH Electrodes
Affordable, high-accuracy pH electrodes for every application
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
Environmental Express/ Oakton high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility.
$170.99 - $462.57CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrode, 12x110 mm, Glass Body, Single Junction, Refill
$462.57CAD / Each
COMPARE
pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill
$170.99CAD / Each
COMPARE
pH Electrode, 12x106 mm, Epoxy Body, Double Junction, Refill
$346.98CAD / Each
COMPARE
pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refillable
$170.99CAD / Each
COMPARE
pH Electrode, 6x106 mm, Epoxy Body, Single Junction, Sealed
$272.28CAD / Each
COMPARE
pH Electrode, 12x106 mm, Epoxy Body, Double Junction, Sealed
$303.67CAD / Each
COMPARE
pH Electrode, 12x110 mm, Glass Body, Double Junction, Refill
$366.28CAD / Each
COMPARE
pH Electrode, 6x106 mm, Glass Body, Double Junction, Refill
$370.19CAD / Each
COMPARE
pH Electrode, 12x110 mm, Epoxy Body, Double Junction, Sealed
$262.26CAD / Each
Showing 1- 9 of 9