Environmental Express® 2000 Series Multiparameter Benchtop Meters
Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring pH/mV, EC, Ion, DO, BOD parameters
  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Meter Kit formats include probes, 35419-54, -60, -65, -74, -78
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level
Environmental Express provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.
These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.
$1,330.47 - $3,195.78CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Meter Type Power (VAC) Description
Availability Pricing
COMPARE
Conductivity/Resistivity/TDS/Salinity/Temp (°C/°F) 100 to 240 EC 2000 Benchtop Meter Kit
$1,616.31CAD / Each
COMPARE
Conductivity/Resistivity/TDS/Salinity/Temp (°C/°F) 100 to 240 EC 2000 Benchtop Meter with Electrode Stand
$1,330.47CAD / Each
COMPARE
pH/ORP/Ion/Conductivity/Resistivity/TDS/Salinity/Temp (°C/°F) 100 to 240 PC 2000 Benchtop Meter Kit
$2,373.63CAD / Each
COMPARE
pH/ORP/Ion/Conductivity/Resistivity/TDS/Salinity/Temp (°C/°F) 100 to 240 PC 2000 Benchtop Meter with Electrode Stand
$1,803.43CAD / Each
COMPARE
pH/ORP/Ion/Temp (°C/°F) 100 to 240 Ion 2000 Benchtop Meter Kit
$1,899.20CAD / Each
COMPARE
pH/ORP/Ion/Temp (°C/°F) 100 to 240 Ion 2000 Benchtop Meter with Electrode Stand
$1,653.14CAD / Each
COMPARE
DO/BOD/Temp (°C/°F) 100 to 240 DO 2000 Benchtop Meter Kit
$2,764.08CAD / Each
COMPARE
DO/BOD/Temp (°C/°F) 100 to 240 DO 2000 Benchtop Meter with Electrode Stand
$1,642.83CAD / Each
COMPARE
pH/ORP/Ion/DO/BOD/Temp (°C/°F) 100 to 240 PD 2000 Benchtop Meter Kit
$3,195.78CAD / Each
COMPARE
pH/ORP/Ion/DO/BOD/Temp (°C/°F) 100 to 240 PD 2000 Benchtop Meter with Electrode Stand
$1,899.20CAD / Each
Showing 1- 10 of 10