Enviro-Genie Benchtop Incubating/Refrigerating Shaker
Five capabilities in one–refrigerate, incubate, rotate, rock, and stir!
  • Features two integrated magnetic stirrers with low/high speed adjustment
  • Precise temperature control and uniformity – range from 4 to 75°C
  • RS-232 port allows for remote operation and data acquisition
Compact multifunctional shaker features a two-sided magnetic platform that easily converts from rocker to rotator to stirrer with a simple adjustment. Microprocessor controls are easy to use with a large LCD displaying speed, temperature, and time. Timer displays elapsed time or remaining time until the programmed shut down time for up to 99 hours. Temperature and time have audible and visible alarms.
Air is continuously circulated throughout the chamber ensuring temperature consistency and uniformity. Environmental conditions are achieved via thermo-electric (Peltier) refrigeration and heating; no compressor is necessary. A shatterproof window makes sample inspection easy without disturbing chamber environment.
Maximize the shakers abilities with a variety of accessories (sold separately). Optional shaker attachment transforms the unit into an orbital shaker in seconds.
$10,195.86 - $10,402.13CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-1200 Enviro- Incubating/Refrigerating Rotating Shaker, 120V. RK-51603-06 Mfr # SI-1200
COMPARE
120
$10,402.13CAD / Each
Scientific Industries Genie SI-1201 Enviro- Incubating/Refrigerating Rotating Shaker, 230V. RK-51603-07 Mfr # SI-1201
COMPARE
230
$10,195.86CAD / Each