This product is now discontinued, View Related Products
Elma® Elmasonic P Ultrasonic Cleaners with Heat and Variable Power
Handle both tough and gentle applications with these dual-frequency cleaning baths
  • Operate at 37 kHz for difficult applications and 80 kHz for sensitive and small items
  • Easily preselect run time and temperature with rotary dials
  • Degas HPLC samples and solvents
  • Obtain uniform sound waves and cleaning from the sweep function
  • Activate the pulse function for 20% increase in ultrasonic power and soil removal
The Elmasonic P series of ultrasonic cleaners come in six different sizes and feature seven convenient operating modes. Normal mode is great for mixing, dissolving, and dispersing samples. Pulse-function increases power 20% for ultra-efficient cleaning—ideal for tough-to-remove pastes and soils. Activate the sweep function to ensure uniform sound and power throughout the entire bath. Degas mode is perfect for quickly removing air from HPLC samples and solvents. The dual-selection power control lets you choose 37 kHz operation for standard and industrial applications, or 80 kHz for smaller or more delicate cleaning needs. The pause function allows for temporary operation interruption, and auto-start mode will begin operation once the preselected temperature is reached.
Select heating temperature from 30 to 80°C, in 5° increments, with a rotary dial. Choose a maximum temperature for sensitive items and instruments—LED light illuminates if reached. Set to operate continuously or in timed runs, from 1 to 30 minutes, also selected with a dial. Safety features include auto-shutoff if running for 12 hours continuously or if bath temperature exceeds 90°C, and a sealed display and elevated feet to protect against splashes and spills.
$1,636.94 - $5,489.85CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Frequency Output (KHz) Power (VAC)
Availability Pricing
Elmasonic P30H Ultrasonic Cleaner with Heat and Variable Power, 0.75 gal.; 120 VAC RK-59988-23 Mfr # 103 3218
COMPARE
0.7 37 or 80 120
$1,711.34CAD / Each
Elmasonic P60H Ultrasonic Cleaner with Heat and Variable Power, 1.5 gal.; 120 VAC RK-59988-24 Mfr # 103 3240
COMPARE
1.5 37 or 80 120
$2,425.94CAD / Each
Elmasonic P70H Ultrasonic Cleaner with Heat and Variable Power, 2 gal.; 120 VAC RK-59988-25 Mfr # 103 3325
COMPARE
1.8 37 or 80 120
$2,515.08CAD / Each
Elmasonic P120H Ultrasonic Cleaner with Heat and Variable Power, 3.5 gal.; 120 VAC RK-59988-26 Mfr # 103 8103
COMPARE
3.4 37 or 80 120
$4,241.15CAD / Each
Elmasonic P180H Ultrasonic Cleaner with Heat and Variable Power, 4.75 gal.; 120 VAC RK-59988-27 Mfr # 103 8158
COMPARE
4.75 37 or 80 120
$4,628.65CAD / Each
Elmasonic P300H Ultrasonic Cleaner with Heat and Variable Power, 7.5 gal.; 120 VAC RK-59988-28 Mfr # 103 8008
COMPARE
7.4 37 or 80 120
$5,208.43CAD / Each
Elmasonic P30H Ultrasonic Cleaner with Heat and Variable Power, 0.75 gal.; 220 VAC RK-59989-23 Mfr # 101 3737
COMPARE
0.7 37 or 80 220
$1,636.94CAD / Each
Elmasonic P60H Ultrasonic Cleaner with Heat and Variable Power, 1.5 gal.; 220 VAC RK-59989-24 Mfr # 101 3761
COMPARE
1.5 37 or 80 220
$2,336.80CAD / Each
Elmasonic P70H Ultrasonic Cleaner with Heat and Variable Power, 2 gal.; 220 VAC RK-59989-25 Mfr # 101 3735
COMPARE
1.8 37 or 80 220
$2,425.94CAD / Each
Elmasonic P120H Ultrasonic Cleaner with Heat and Variable Power, 3.5 gal.; 220 VAC RK-59989-26 Mfr # 101 3777
COMPARE
3.4 37 or 80 220
$4,107.81CAD / Each
Elmasonic P180H Ultrasonic Cleaner with Heat and Variable Power, 4.75 gal.; 220 VAC RK-59989-27 Mfr # 101 3804
COMPARE
4.75 37 or 80 220
$5,489.85CAD / Each
Elmasonic P300H Ultrasonic Cleaner with Heat and Variable Power, 7.5 gal.; 220 VAC RK-59989-28 Mfr # 101 3810
COMPARE
7.4 37 or 80 220
$5,059.62CAD / Each
Showing 1- 12 of 12