Cole-Parmer® HM-200 Series Multiple Volume Heating Mantles
Accepts 250 to 1000-mL round-bottom flasks saving you money
  • Maximum element temperature of 842°F (450°C)
  • Cool to the touch offers safe handling
  • Built-in energy regulator
Flexible coiled heating element wraps the heater around round-bottom flasks to optimize heat transfer. The flask does not come in direct contact with the heating element via a grounded earth screen plus double fuse protection provides maximum safety. Control temperature with a built-in 1 to 10 scale power controller. Powder-coated aluminum housing makes it easy to clean. Two colored lights indicate "power on" and "heat on."
$1,057.16 - $1,060.84CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Max temperature (° F) Power (VAC)
Availability Pricing
COMPARE
1000 842 230
$1,060.84CAD / Each
COMPARE
1000 842 115
$1,057.16CAD / Each