Cole-Parmer® HT-200 Series Grounded Heating Tapes
Flexible heaters—fit metal pipes and glassware
  • Maximum element temperature of 842°F (450°C)
  • Braided earth wire beneath the outer sheath
  • Approximately 25 mm wide
The flexible fiberglass heating tapes are ideal for heating columns, pipes, valves, and transfer lines, where applications need to be observed. The heating element and braided ground wire are wound on a fiberglass core and covered in braided fiberglass outer sleeve. Tapes are not water-resistant; for use in dry conditions only. Tapes are supplied with 3.5" (9 cm) insulated unheated flexible leads at each end. Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
$614.40 - $4,169.69CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
COMPARE
2 842 230
$614.40CAD / Each
COMPARE
2 842 115
$646.82CAD / Each
COMPARE
3 842 230
$626.19CAD / Each
COMPARE
3 842 115
$614.40CAD / Each
COMPARE
4 842 230
$649.76CAD / Each
COMPARE
4 842 115
$671.87CAD / Each
COMPARE
6 842 230
$1,022.53CAD / Each
COMPARE
6 842 115
$761.74CAD / Each
COMPARE
8 842 230
$850.15CAD / Each
COMPARE
8 842 115
$917.92CAD / Each
COMPARE
12 842 230
$1,583.89CAD / Each
COMPARE
12 842 115
$1,642.83CAD / Each
COMPARE
16 842 230
$1,427.71CAD / Each
COMPARE
16 842 115
$1,326.05CAD / Each
COMPARE
24 842 230
$2,909.95CAD / Each
COMPARE
32 842 230
$4,169.69CAD / Each
Showing 1- 16 of 16