Cole-Parmer® HM-200 Series Flask/Funnel Heating Mantles
Opening in base accommodates a wide variety of flasks and 60º funnels
  • Ceramic fiber insulation keeps case cool
  • Built-in power controller offers convenience
  • Chemical-resistant polypropylene housing
Funnel-shaped cavity with a base opening allows uses with 60° funnels and a wide variety of flasks. Heating mantles have maximum element temperature of 842°F(450°C). Ceramic fiber insulation keeps the polypropylene housing cool. Built-in power controller with dial markings from 1 to 10 offer repeatability. The "power on" and "heater on" lights indicate if the unit is on.
The 36101-series heating mantles with stainless steel (SS) liner over the heating element offer protection against spills and make cleaning easier. The 36104-series heating mantles include a safety earth (ground) screen to provide operator safety. All mantles (except 5000-mL models) feature a single support rod clamp that accepts a 1/2" rod.
$785.32 - $2,018.54CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC) Description
Availability Pricing
COMPARE
1000 230 Flask/Funnel Heating Mantle with SS liner, 50 to 1000 mL; 230 VAC
$1,259.75CAD / Each
COMPARE
5000 115 Flask/Funnel Heating Mantle with SS liner, 500 to 5000 mL; 115 VAC
$2,018.54CAD / Each
COMPARE
5000 230 Flask/Funnel Heating Mantle with SS liner, 2000 to 5000 mL; 230 VAC
$2,018.54CAD / Each
COMPARE
50 115 Flask/Funnel Heating Mantle, 10 to 50 mL; 115 VAC
$785.32CAD / Each
COMPARE
50 230 Flask/Funnel Heating Mantle, 10 to 50 mL; 230 VAC
$785.32CAD / Each
COMPARE
250 230 Flask/Funnel Heating Mantle, 100 to 250 mL; 230 VAC
$897.29CAD / Each
COMPARE
1000 115 Flask/Funnel Heating Mantle, 500 to 1000 mL; 115 VAC
$881.09CAD / Each
COMPARE
1000 230 Flask/Funnel Heating Mantle, 500 to 1000 mL; 230 VAC
$948.13CAD / Each
COMPARE
5000 115 Flask/Funnel Heating Mantle, 2000 to 5000 mL; 115 VAC
$1,407.09CAD / Each
COMPARE
5000 230 Flask/Funnel Heating Mantle, 2000 to 5000 mL; 230 VAC
$1,623.68CAD / Each
Showing 1- 10 of 10