Cole-Parmer® HM-200 Series Controlled Aluminum Heating Mantles
Don't get burned—outside stays "safe to the touch"
  • Hold round-bottom, Erlenmeyer, and volumetric flasks
  • Maximum element temperature of 842°F (450°C)
  • Built-in energy regulator
Safely heat round-bottom flasks to 842°F (450°C). Mantles have built-in power controller with a 1 to 10 scale for easy repeatability. Two neon lights indicate "power on" and "heat on." The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Powder-coated aluminum outer casing stays cool and is easy to clean. A stainless steel grounded earth screen and double fuses provided added safety. All heating mantles feature a support bracket for holding a support rod.
$800.05 - $1,001.17CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC) Power (watts)
Availability Pricing
COMPARE
100 115 100
$800.05CAD / Each
COMPARE
100 230 100
$803.00CAD / Each
COMPARE
250 115 200
$881.09CAD / Each
COMPARE
250 230 200
$885.51CAD / Each
COMPARE
500 115 280
$1,001.17CAD / Each
COMPARE
500 230 280
$998.22CAD / Each
COMPARE
1000 115 380
$998.22CAD / Each
COMPARE
1000 230 380
$984.22CAD / Each
Showing 1- 8 of 8