This product is now discontinued, View Related Products
Economical Noise Dosimeters 7764
Ideal for employee noise-exposure monitoring
Meters display instantaneous or accumulated % dose, 8-hour dose, dB level, exposure time, and peak events for efficient noise-exposure monitoring. Lightweight for workers comfort. Select model with RS-232 output for downloading your data.
$1,060.83 - $5,937.76CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Decibel (Db) High Range (dB) Decibel (Db) Low Range (dB) Meter Class
Availability Pricing
Extech 407355 Economical Noise Dosimeter with RS-232 RK-01621-10 Mfr # 407355
COMPARE
140 70
$1,060.83CAD / Each
Extech 407355-KIT-5 Data Logging Noise Dosimeter Kit RK-01621-11 Mfr # 407355-KIT-5
COMPARE
140 70
$5,937.76CAD / Each