This product is now discontinued, View Related Products
Economical Digital Shaking Water Baths
  • Adjustable temperature, stroke length, and speed give these 10- and 20-liter baths the versatility you want
These digital shaking water baths feature a microprocessor-based keypad and digital display with preset time, continuous run, rpm, and temperature. Nonvolatile memory safely stores your instructions during a power outage. Slow-start and stop feature deters splashing and prevents sample loss. Low-water level signals heater shutoff for added sample safety.
Enamel-coated aluminum housing, and seamless, corrosion resistant stainless steel chamber with removable basket are easy to maintain. Secure flasks in the basket with industry-standard flask clamps sold separately. Optional gable-type cover (not included) maintains temperature accuracy — sold separately. Adjustable springs that stretch across the basket to hold racks and other objects in place while the basket shakes are also optional and sold seperately.
$4,199.16 - $5,472.17CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
EDVOTEK Economical Digital Shaking Water Bath, 10 L capacity, 110 VAC RK-12052-00 Mfr # 5027-110V
COMPARE
115 10
$4,199.16CAD / Each
EDVOTEK Economical Digital Shaking Water Bath, 10 L capacity, 220 VAC RK-12052-05 Mfr # 95027
COMPARE
230 10
$4,431.96CAD / Each
EDVOTEK Economical Digital Shaking Water Bath, 20 L capacity, 110 VAC RK-12052-10 Mfr # SHWB20-110V
COMPARE
115 20
$5,472.17CAD / Each
EDVOTEK Economical Digital Shaking Water Bath, 20 L capacity, 220 VAC RK-12052-15 Mfr # SHWB20-220V
COMPARE
230 20
$5,472.17CAD / Each