This product is now discontinued, View Related Products
Ebro TTX Compact Foodservice Thermocouple Thermometers
Ideal for use in the food industry
  • IP55-rated waterproof case
These thermometers are constructed of ABS plastic and have an IP55 rating producing a waterproof and shock-resistant product ideal for use in the food industry. The use of an advanced internal microprocessor that updates every five seconds and a highly accurate type T thermocouple produces an accuracy of ±0.8%. The product features a Hold function, °F/°C selectable units, and auto power-off after 15 seconds of inactivity to save on battery power.
$117.87 - $173.86CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Data Logging Max Temperature (° F) Types Of Probes Accepted
Availability Pricing
Ebro 1340-5100 TFE 100 Compact Food Thermometer with Remote Probe RK-91210-35 Mfr # 1340-5100
COMPARE
660 Type T
$173.86CAD / Each
Ebro 1340-5110 TFE110 Compact Food Thermometer with Attached Probe RK-91210-36 Mfr # 1340-5110
COMPARE
660 Type T
$117.87CAD / Each