This product is now discontinued, View Related Products
EBRO TFX Compact Waterproof Digital RTD Thermometers
Ideal for use in harsh environments like labs, production and the food industry
  • IP67-rated housing and key pad
  • Compact size allows for convenient storage
These meters feature a robust splash/impact-resistant ABS housing, corrosion-resistant, stainless steel probe, battery life indicator and an auto shutoff. A large LCD displays the temperature readings and is dishwasher resistant. Multifunction models have max/min and hold capabilities. Available with a choice of probes that are per installed; blunt-tip for general-purpose use, penetration for semi soft materials and a remote sensor with perpetrating end for increased flexibility
$493.59 - $1,038.74CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Ebro 1340-5417 IP-67 RTD Thermometer with Cert of Conformance, Blunt Tip; -58 to 572F RK-90111-10 Mfr # 1340-5417
COMPARE
572 RTD 1
$493.59CAD / Each
Ebro 1340-5418 IP-67 RTD Thermometer with Cert of Conformance, Sharp Tip; -58 to 572F RK-90111-11 Mfr # 1340-5418
COMPARE
572 RTD 1
$500.95CAD / Each
Ebro 1340-5416 IP-67 RTD Thermometer with Cert of Conformance, Remote Sharp Tip; -58 to 572F RK-90111-12 Mfr # 1340-5416
COMPARE
572 RTD 1
$493.59CAD / Each
Ebro 1340-5437 IP-67 RTD Thermometer with Cert of Conformance, Blunt Tip; -148 to 932F RK-90111-13 Mfr # 1340-5437
COMPARE
932 RTD 1
$1,038.74CAD / Each
Ebro 1340-5438 IP-67 RTD Thermometer with Cert of Conformance, Sharp Tip; -148 to 932F RK-90111-14 Mfr # 1340-5438
COMPARE
932 RTD 1
$1,038.74CAD / Each
Ebro 1340-4268 IP-67 RTD Thermometer with Cert of Conformance, Remote Sharp Tip; -148 to 932F RK-90111-15 Mfr # 1340-4268
COMPARE
752 RTD 1
$633.56CAD / Each