This product is now discontinued, View Related Products
Dyna-Lume LED Illuminators
High intensity light for your inspection applications
  • Fully posable goosenecks allow for multiple light configurations
Order the optional magnifier for convenient, close-up inspections.
$446.44 - $669.66CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bulb Type Power (VAC) Description
Availability Pricing
Scientific Instruments 240-361 LED Illuminator, Single head, Narrow beam RK-09809-20 Mfr # 240-361
COMPARE
Narrow/15º 100 to 240 LED Illuminator, Single head, Narrow beam
$446.44CAD / Each
Scientific Instruments 240-362 LED Illuminator, Dual head, Narrow beam RK-09809-25 Mfr # 240-362
COMPARE
Narrow/15º 100 to 240 LED Illuminator, Dual head, Narrow beam
$589.72CAD / Each
Scientific Instruments 240-363 LED Illuminator, Single head, Combination beam RK-09809-30 Mfr # 240-363
COMPARE
Combination/24º 100 to 240 LED Illuminator, Single head, Combination beam
$514.21CAD / Each
Scientific Instruments 240-364 LED Illuminator, Dual head, Combination beam RK-09809-35 Mfr # 240-364
COMPARE
Combination/24º 100 to 240 LED Illuminator, Dual head, Combination beam
$669.66CAD / Each