This product is now discontinued, View Related Products
Dyna-Lume Clip-On Illuminators
Clip-on design allows illumination almost anywhere
  • Plastic ring guard provides safe fingertip adjustment
These quartz halogen illuminators securely clamp onto any horizontal, vertical, or round surface edge up to 3⁄4" dia. Self-supporting, 8"L flexible arm and swivel-mounted head allow you to position the light beam exactly where you need it.
Low-voltage power supply is connected to head assembly via a 10-ft cable. Intensity control dial lets you precisely adjust light levels. Miniature quartz halogen bulb (included) measures only 1-3⁄8" in diameter — delivering cool shadowless illumination (3000K color temperature). Order optional infrared heat shield 09809-96 for close-up work on heat-sensitive samples.
What’s included: remote power supply, halogen bulb(s), and 5-ft power cord.
$343.15 - $477.38CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bulb Type Power (VAC) Description
Availability Pricing
Scientific Instruments 240-341 Clip-On Illuminator, Single-Head RK-41600-00 Mfr # 240-341
COMPARE
Halogen 120 Clip-On Illuminator, Single-Head
$343.15CAD / Each
Scientific Instruments 240-342 Clip-On Illuminator, Dual-Head RK-41600-10 Mfr # 240-342
COMPARE
Halogen 120 Clip-On Illuminator, Dual-Head
$477.38CAD / Each