This product is now discontinued, View Related Products

Dwyer Differential Pressure Wet-Wet Gauge with Transmitter

The series 631B expands on the Capsuhelic Differential Pressure gauge and adds an integral transmitter output. The high pressure limits permit the use of the gauge /transmitter on system pressures of up to 500 psig while allowing differentials to be read at less than 0.1” w.c. This model has a brass body and is compatible with air and compatible liquids. The unit also adds push button calibration along with a NEMA 4X housing.

APPLICATIONS lbr>
Building automation systems lbr>
Chillers lbr>
Filter monitoring lbr>
$1,373.20 - $1,421.82CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer 631B-0 Capsuhelic Differential Pressure Transmitter0.5" WC RK-68457-40 Mfr # 631B-0
COMPARE
±3% full-scale 0.1\" WC
$1,373.20CAD / Each
Dwyer 631B-1 Capsuhelic Differential Pressure Transmitter1" WC RK-68457-41 Mfr # 631B-1
COMPARE
±3% full-scale 0.1\" WC
$1,373.20CAD / Each
Dwyer 631B-2 Capsuhelic Differential Pressure Transmitter, 2" WC RK-68457-42 Mfr # 631B-2
COMPARE
±3% full-scale 0.1\" WC
$1,373.20CAD / Each
Dwyer 631B-3 Capsuhelic Differential Pressure Transmitter, 5" WC RK-68457-43 Mfr # 631B-3
COMPARE
±3% full-scale 0.1\" WC
$1,421.82CAD / Each
Dwyer 631B-5 Capsuhelic Differential Pressure Transmitter, 25" WC RK-68457-44 Mfr # 631B-5
COMPARE
±3% full-scale 0.1\" WC
$1,421.82CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat