This product is now discontinued, View Related Products
Dwyer DH-3 Differential Pressure Controller
Digital gauge combines control relay switches and a transmitter with current output
  • Allows the reduction of several instruments with one product
  • Save inventory, installation time, and money
These controllers are an ideal instrument for pressure, velocity and flow applications. The differential pressure controller allows the selection of pressure, velocity or volumetric flow operation in common engineering units. Digital gauge features two spdt relays with adjustable deadband and are provided along with a scalable 4-20 mA process output.
The gauge’s keypad allow you to access 5 simplified menus that provide access to: security level; selection of pressure, velocity or flow operation; selection of engineering units; K-factor for use with flow sensors; rectangular or circular duct for inputting area in flow applications; set point control or set point and alarm operation; alarm operation as a high, low or high/low alarm; automatic or manual alarm reset; alarm delay; view peak and valley process reading; digital damping for smoothing erratic process applications; scaling the 4-20 mA process output to fit your applications range and field calibration.
Applications
Airflow in ducts
Filter status
Static pressures in ducts or buildings
Damper control
Fan control
$440.54 - $657.57CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
Dwyer DH3-002 Differential Pressure Controller, range 0-0.25" WC RK-68063-00 Mfr # DH3-002
COMPARE
±1% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$657.57CAD / Each
Dwyer DH3-003 Differential Pressure Controller, range 0-0.5" WC RK-68063-02 Mfr # DH3-003
COMPARE
±1%
$440.54CAD / Each
Dwyer DH3-004 Differential Pressure Controller, range 0-1" WC RK-68063-04 Mfr # DH3-004
COMPARE
±1%
$440.54CAD / Each
Dwyer DH3-005 Differential Pressure Controller, range 0-2.5" WC RK-68063-06 Mfr # DH3-005
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$440.54CAD / Each
Dwyer DH3-006 Differential Pressure Controller, range 0-5" WC RK-68063-08 Mfr # DH3-006
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$440.54CAD / Each
Dwyer DH3-007 Differential Pressure Controller, range 0-10" WC RK-68063-10 Mfr # DH3-007
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$440.54CAD / Each
Dwyer DH3-009 Differential Pressure Controller, range 0-25" WC RK-68063-12 Mfr # DH3-009
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$440.54CAD / Each
Dwyer DH3-010 Differential Pressure Controller, range 0-50" WC RK-68063-14 Mfr # DH3-010
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$535.58CAD / Each
Dwyer DH3-011 Differential Pressure Controller, range 0-100" WC RK-68063-16 Mfr # DH3-011
COMPARE
±0.5% at 77°F (25°C) including hysteresis and repeatability (after 1 hour warm-up)
$440.54CAD / Each
Showing 1- 9 of 9