This product is now discontinued, View Related Products
Dwyer Capsuhelic Differential Pressure Gauges
For air or compatible gases or liquids. Measures pressure, vacuum or differential. Suitable for internal pressures o 500 psig. May be flush or surface mounted. 5'' diameter, requires a 4-13/16'' hole for flush mounting.
$614.11 - $941.50CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer 4000-0 Capshelic 0/.5 Inh2O Differential P Gauge RK-68462-33 Mfr # 4000-0
COMPARE
±3% full-scale 0.01
$722.40CAD / Each
Dwyer 4002 Capsuhelic 0/2.0 Inh2O Differential P Gauge RK-68462-34 Mfr # 4002
COMPARE
±3% full-scale 0.05
$722.40CAD / Each
Dwyer 4005 Capsuhelical0/5.0inH2O Differential P Gauge RK-68462-35 Mfr # 4005
COMPARE
±3% full-scale 0.10
$722.40CAD / Each
Dwyer 4010 Capsuhelic 0/10 Inh2O Differential P Gauge RK-68462-36 Mfr # 4010
COMPARE
±3% full-scale 0.20
$722.40CAD / Each
Dwyer 4020 Capsuhelic 0/20 Inh2O Differential P Gauge RK-68462-37 Mfr # 4020
COMPARE
±3% full-scale 0.50
$614.11CAD / Each
Dwyer 4025 Capsuhelic 0/25 Inh2O Differential P Gauge RK-68462-38 Mfr # 4025
COMPARE
±3% full-scale 0.50
$722.40CAD / Each
Dwyer 4100 Capsuhelic 0/100 Inh2O Differential P Gauge RK-68462-39 Mfr # 4100
COMPARE
±3% full-scale 2
$722.40CAD / Each
Dwyer 4200 Capsuhelic 0/200 Inh2O Differential P Gauge RK-68462-40 Mfr # 4200
COMPARE
±3% full-scale 5
$722.40CAD / Each
Dwyer 4500 Capsuhelic 0/500 Inh2O Differential P Gauge RK-68462-41 Mfr # 4500
COMPARE
±3% full-scale 10
$722.40CAD / Each
Dwyer 4201 Capsuhelic 0/1PSI Differential P Gauge RK-68462-42 Mfr # 4201
COMPARE
±3% full-scale 0.02
$696.91CAD / Each
Dwyer 4205 Capsuhelic 0/5PSI Differential P Gauge RK-68462-43 Mfr # 4205
COMPARE
±3% full-scale 0.10
$722.40CAD / Each
Dwyer 4215 Capsuhelic 0/15PSI Differential P Gauge RK-68462-44 Mfr # 4215
COMPARE
±3% full-scale 0.50
$722.40CAD / Each
Dwyer 4260S Capsuhelic 0/60PSI Differential P Gauge RK-68462-45 Mfr # 4260S
COMPARE
60 2
$941.50CAD / Each
Dwyer 4040 Capsuhelic 0/40 Inh2O Differential P Gauge RK-68462-48 Mfr # 4040
COMPARE
±3% full-scale 1
$614.11CAD / Each
Dwyer 4230S Capsuhelic 0/30PSI Differential P Gauge RK-68462-49 Mfr # 4230S
COMPARE
30 1
$941.50CAD / Each
Showing 1- 15 of 15