DWK Wheaton® Potter-Elvehjem Tissue Grinders
Compatible with overhead stirrers, perfect for frequent sample processing
  • PTFE-tipped stainless steel rod fits overhead stirrer collets or chucks designed for 0.25" shafts
These grinders come with a PTFE pestle for use with soft tissue. Stainless steel rod is 0.25" (6.3 mm) in diameter. Grinding chamber clearance is 0.004" to 0.006". Unit can be autoclaved.
$406.66 - $411.08CAD / Case of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (mm)
Availability Pricing
DWK Life Sciences (Wheaton) 358029 Potter-Elvehjem Tissue Grinders, Glass, 2 mL; 2/Cs RK-04468-14 Mfr # 358029
COMPARE
2 203
$406.66CAD / Case of 2
DWK Life Sciences (Wheaton) 358034 Potter-Elvehjem Tissue Grinders, Glass, 5 mL; 2/Cs RK-04468-16 Mfr # 358034
COMPARE
5 219
$411.08CAD / Case of 2