This product is now discontinued, View Related Products
Dupont Tyvek Cleanroom Coveralls, 25/case
Tyvek® IsoClean® Coveralls are made using an exclusive flash spinning process. Excellent inherent barrier to dry particles, microorgainisms, and nonhazardous liquids. Coveralls feature zipper front, bound seams, and elastic wrists and ankles. White.
Compliance: ISO Class 5
$493.59 - $599.38CAD / Case of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Size
Availability Pricing
DuPont Tyvek IsoClean Coveralls, small, cleaned and individually wrapped RK-33684-00 Mfr # IC182BWHSM00250C
COMPARE
Small
$559.59CAD / Case of 25
DuPont Tyvek cleanroom coveralls; medium, 25/cs RK-33684-03 Mfr # IC182BWHMD00250C
COMPARE
Medium
$559.59CAD / Case of 25
DuPont cleanroom coveralls; large, 25/cs RK-33684-04 Mfr # IC148BWHLG00250C
COMPARE
Large
$493.59CAD / Case
DuPont Tyvek cleanroom coveralls; large, 25/cs RK-33684-05 Mfr # IC182BWHLG00250C
COMPARE
Large
$594.81CAD / Case of 25
DuPont cleanroom coveralls; XL, 25/cs RK-33684-06 Mfr # IC148BWHXL00250C
COMPARE
X-Large
$493.59CAD / Case
DuPont IC182BWH00250C Coveralls, x-large, cleaned and individually wrapped RK-33684-07 Mfr # IC182BWHXL00250C
COMPARE
X-Large
$599.38CAD / Case of 25