This product is now discontinued, View Related Products
Dupont Cleanroom Coveralls
Pro/Clean® Coveralls are made from a unique breathable composite material for critical environments. This fabric provides excellent particulate and light splash protection. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist. White.
Compliance: ISO Class 6, 7, and 8 (formerly Class 1000, 10,000, and 100,000)
$308.23 - $317.37CAD / Case of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Size
Availability Pricing
DuPont Pro/Clean Coveralls, medium, bulk packed RK-33684-26 Mfr # PC143SWHMD00250B
COMPARE
White Medium
$308.23CAD / Case of 25
DuPont Pro/Clean Coveralls, large, bulk packed RK-33684-27 Mfr # PC143SWHLG00250B
COMPARE
White Large
$308.23CAD / Case of 25
DuPont PC143SWH00250B Coveralls, x-large, bulk packed RK-33684-28 Mfr # PC143SWHXL00250B
COMPARE
White X-Large
$308.23CAD / Case of 25
DuPont Pro/Clean Coveralls, xx-large, bulk packed RK-33684-29 Mfr # PC143SWH2X00250B
COMPARE
White XX-Large
$317.37CAD / Case of 25