This product is now discontinued, View Related Products

Drummond Original Pipet-Aid Pipette Controllers

Dedicated workstation pipetting device
  • Can be used with most (standard) glass or plastic pipettes
Electrically-powered pipetting device is a convenient alternative to use bulb aspirators. Easy to use volume control mechanism allows precise fluid intake and delivery. A check valve protects against accidental fluid intake in the handle.
$427.28 - $528.95CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Speed Description
Availability Pricing
Drummond 4-000-110 Pipette Controller, 5 mL/sec, 110 VAC RK-14400-00 Mfr # 4-000-110
COMPARE
5000 5.0mL/sec Pipette Controller, 5 mL/sec, 110 VAC
$427.28CAD / Each
Drummond 4-000-110-TC Pipette Controller, 5 mL/sec, Tissue-Culture; 110V RK-14400-02 Mfr # 4-000-110-TC
COMPARE
5000 5.0mL/sec Pipette Controller, 5 mL/sec, Tissue-Culture; 110V
$477.38CAD / Each
Drummond 4-000-220 Pipette Controller, 5 mL/sec; 220 VAC RK-14400-04 Mfr # 4-000-220
COMPARE
5000 5.0mL/sec Pipette Controller, 5 mL/sec; 220 VAC
$477.38CAD / Each
Drummond 4-000-220-TC Pipette Controller, 5 mL/sec, Tissue-Culture; 220V RK-14400-06 Mfr # 4-000-220-TC
COMPARE
5000 5.0mL/sec Pipette Controller, 5 mL/sec, Tissue-Culture; 220V
$528.95CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat