This product is now discontinued, View Related Products
Drum and Barrel Top-to-Bottom Electric Motor Tube Mixers
Top-to-bottom movement provides thorough mixing without creating a vortex
These efficient tube mixers allow you to thoroughly mix the contents of 55-gallon (200 liter) drums without removing the drum lid. The 40" (1016 mm) long mixing tube fits into standard 2" (51 mm) bungs and generates a powerful top-to-bottom flow.
A shaft-mounted turbine inside the mixing tube creates a steady suction effect at four upper intake ports. Mixers draw approximately 13 to 14 gallons (49 to 53 liters) of liquid (750 cp max) per minute through the 2" (51 mm) OD mixing tube. The forcefully expelled liquid resuspends and dissolves settled particles. Choose either an electric or air-drive motor mixer.
Electric motor mixers have 1/4-hp motors that maintain a 3450-rpm mixing speed and operate on 115 VAC, 60 Hz. Mixers feature an on/off switch on the motor for your convenience. Select either a totally enclosed, fan-cooled (TEFC) motor or CSA-approved explosion-proof motor (rated for Class 1, Division 1, Group D conditions).
$2,949.73 - $3,145.69CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Motor hp
Availability Pricing
Finish Thompson Drum and Barrel Top-to-Bottom Electric Motor Tube Mixer, 115V RK-04316-20 Mfr # A101134 + DTMS007-CP
COMPARE
3450 3450 1/4
$3,145.69CAD / Each
Finish Thompson Drum/barrel tube mixer with an explosion-proof motor, 115 VAC RK-04316-24 Mfr # A101135 & DTMS007
COMPARE
3450 3450 1/4
$2,949.73CAD / Each