Disruptor Genie
Enhanced vortex action for maximum high-speed mixing and agitating
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
Disruptor Genie® Shaker is ideal for use in cold rooms or incubators. Disruptor features dramatically increased disruption efficiency - ideal for difficult cell disruptions/homogenizations. Simple to use-just set the timer for desired run time. The multidirectional action simultaneously agitates and vortexes at high-speed. The single high-speed agitation eliminates operator variability.
$1,185.34 - $1,281.11CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-D238 Disruptor Shaker with Timer, 3000 rpm, 120 VAC RK-04724-31 Mfr # SI-D238
COMPARE
120
$1,281.11CAD / Each
Scientific Industries Genie SI-D248 Disruptor Shaker with Timer, 2850 rpm, 230 VAC RK-04724-36 Mfr # SI-D248
COMPARE
230
$1,185.34CAD / Each