This product is now discontinued, View Related Products
Disposable Sterile Culture Tubes with two-position PE caps
Heavy-walled tubes resist breakage. Choose from clear polystyrene (PS) or natural polypropylene (PP). The 12 x 75 mm tubes have a volume of 6 mL; the 17 x 100 mm tubes have a volume of 16 mL. Tubes are sterile; include two-position PE caps.
$215.11 - $335.93CAD / Box of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Length (mm) Material
Availability Pricing
COMPARE
12 75 PE
$335.93CAD / Box of 1000
COMPARE
12 75 PE
$290.99CAD / Box of 1000
COMPARE
17 100 PE
$225.43CAD / Pkg of 500
COMPARE
17 100 PE
$215.11CAD / Pkg of 500