This product is now discontinued, View Related Products
Disposable Single-Chamber Sampling Lance for Powders and Granulate
Three-zone sampling in one easy step
  • Three spaced openings along shaft sample different layers in one motion
  • Free-flowing powders and small-grain granulates work best
  • Rotating handle closes sampler for transport
  • Gamma sterilized and nonsterile versions available
  • Individually packaged
This HDPE powder and granulate sampler provides a fast all-in-one solution to layer sampling. With three openings spaced apart along the hollow shaft, the product falls into the single interior chamber from three distinct locations simultaneously. No need to repeat the sampling process multiple times. The handle is closed and when rotated, the openings are closed off providing a secure container for transport.
$461.32 - $624.42CAD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Volume (mL)
Availability Pricing
Burkle Disposable Zone Sampling Lance, Closed-Tip, HDPE, 50 cm x 2.5 cm; 100 mL RK-06043-15 Mfr # 5393-4420
COMPARE
19.625 No 100
$461.32CAD / Pkg of 20
Burkle Disposable Zone Sampling Lance, Closed-Tip, HDPE, 50 cm x 2.5 cm, Sterile; 100 mL RK-06043-16 Mfr # 5393-4421
COMPARE
19.625 Yes 100
$522.02CAD / Pkg of 20
Burkle Disposable Zone Sampling Lance, Closed-Tip, HDPE, 100 cm x 2.5 m; 200 mL RK-06043-17 Mfr # 5393-4430
COMPARE
39.375 No 200
$529.09CAD / Pkg of 20
Burkle Disposable Zone Sampling Lance, Closed-Tip, HDPE, 100 cm x 2.5 cm, Sterile; 200 mL RK-06043-18 Mfr # 5393-4431
COMPARE
39.375 Yes 200
$624.42CAD / Pkg of 20