This product is now discontinued, View Related Products
Disposable Liquid Siphon with Draw Handle
Quickly sample low viscosity to slightly viscous liquids
  • Individually packaged
  • Cleanroom manufactured
  • Ergonomic handle and draw bar make it easy to draw samples
  • Gamma sterilized and nonsterile versions available
These HDPE liquid samplers feature an ergonomic handle and a sturdy draw bar to make sample drawing and dispensing a breeze. Samplers are manufactured in a cleanroom and individually packaged to prevent contamination ensuring more accurate results.
$301.68 - $492.85CAD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Burkle Disposable liquid siphon with draw handle, HDPE, 500 mm x 21 mm; 100 mL RK-06043-00 Mfr # 5393-1120
COMPARE
Disposable Liquid Siphon with Draw Handle, 100 mL
$301.68CAD / Pkg of 20
Burkle Disposable liquid siphon with draw handle, HDPE, 500 mm x 21 mm, sterile; 100 mL RK-06043-02 Mfr # 5393-1121
COMPARE
Disposable Liquid Siphon with Draw Handle, 100 mL, Sterile
$356.56CAD / Pkg of 20
Burkle Disposable liquid siphon with draw handle, HDPE, 1000 mm x 21 mm; 200 mL RK-06043-03 Mfr # 5393-1130
COMPARE
Disposable Liquid Siphon with Draw Handle, 200 mL
$370.19CAD / Pkg of 20
Burkle Disposable liquid siphon with draw handle, HDPE, 1000 mm x 21 mm, sterile; 200 mL RK-06043-04 Mfr # 5393-1131
COMPARE
Disposable Liquid Siphon with Draw Handle, 200 mL, Sterile
$492.85CAD / Pkg of 20