Disposable In-Line Gas/Liquid Filters
Choose opaque KYNAR® PVDF for the best chemical compatibility or transparent nylon for lower cost
  • Max differential pressure for all filters is 40 psi (60 psi for 0.01 µm units)
  • Max temperature at 0 psi: nylon is 230°F; stainless steel & KYNAR PVDF are 250°F
Filter gases or liquids with these microfiber filters. All models have a borosilicate glass filter membrane. See table below for applications and efficiency ratings. Liquid efficiency ratings are 98% of retention of particles of noted size; gas efficiency ratings are percentage retention of 0.01 µm particles.
$134.37 - $1,809.32CAD / Pkg of 5
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Housing Material Description
Availability Pricing
COMPARE
PVDF In-Line Gas/Liquid Filters, 93%/25 um, PVDF, 1/4\" OD; 5/Pk
$592.30CAD / Pkg of 5
COMPARE
PVDF In-Line Gas/Liquid Filters, 98%/8 um, PVDF, 1/4\" OD; 5/Pk
$518.26CAD / Pkg of 5
COMPARE
PVDF In-Line Gas/Liquid Filters, 99.99%/2 um, PVDF, 1/4\" OD; 5/Pk
$618.09CAD / Pkg of 5
COMPARE
Nylon Disposable In-Line Gas/Liquid Filters; Nylon w/ carbon granules, 99% gas, 10/pk
$1,000.43CAD / Pkg of 10
COMPARE
Nylon Disposable In-Line Gas/Liquid Filters; Nylon w/ Na- & Ca-OH, 99% gas, 10/pk
$926.76CAD / Pkg of 10
COMPARE
PVDF In-Line Gas/Liquid Filters, 99.9999+%/0.9 um, PVDF, 1/4\" OD; 5/Pk
$546.26CAD / Pkg of 5
COMPARE
PVDF Disposable In-Line Gas/Liquid Filters; PVDF; liquid efficiency, 0.3µ; 5/pack
$749.22CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 93%/25 um, Nylon, 1/4\" OD; 5/Pk
$345.14CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 93%/25 um, Nylon, 1/4 to 3/8\" ID; 10/Pk
$676.29CAD / Pkg of 10
COMPARE
Nylon In-Line Gas/Liquid Filters, 93%/25 um, Nylon, 1/2\" OD; 1/Pk
$136.44CAD / Pkg of 1
COMPARE
Nylon Disposable In-Line Gas/Liquid Filters; Nylon; liquid efficiency, 8µ; 5/pack
$389.34CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.99%/2 um, Nylon, 1/4\" OD; 5/Pk
$424.34CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.99%/2 um, Nylon, 1/4 to 3/8\" ID; 10/Pk
$697.28CAD / Pkg of 10
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.99%/2 um, Nylon, 1/2\" OD; 1/Pk
$134.37CAD / Pkg of 1
COMPARE
Nylon Disposable In-Line Gas/Liquid Filters; Nylon, 99.99% gas, 2 um liquid, 5/pk
$412.18CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.9999+%/0.9 um, Nylon, 1/4\" OD; 5/Pk
$416.60CAD / Pkg of 5
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.9999+%/0.9 um, Nylon, 1/4 to 3/8\" ID; 10/Pk
$661.92CAD / Pkg of 10
COMPARE
Nylon In-Line Gas/Liquid Filters, 99.9999+%/0.3 um, Nylon, 1/4\" OD; 5/Pk
$526.74CAD / Pkg of 5
COMPARE
Nylon Disposable In-Line Gas/Liquid Filters; Nylon; gas efficiency; 100% at 0.01µ
$1,809.32CAD / Pkg of 5
Showing 1- 19 of 19