Disposable Griffin Polypropylene Low-Form Beakers
Molded-in graduations won’t wear off
  • Won’t stick together—stacking rib for easier separation
  • Autoclavable
These disposable polypropylene (PP) beakers are graduated in mL and oz, so there is no need to convert volumes. Molded-in stacking ribs prevent beakers from sticking together when stacked.
$37.42 - $1,156.61CAD / Case of 100
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
15 5 Polypropylene(PP)
$37.42CAD / Case of 100
COMPARE
15 5 Polypropylene(PP)
$346.25CAD / Case of 1000
COMPARE
30 5 Polypropylene(PP)
$44.20CAD / Case of 100
COMPARE
30 5 Polypropylene(PP)
$427.28CAD / Case of 1000
COMPARE
50 5 Polypropylene(PP)
$42.73CAD / Case of 100
COMPARE
50 5 Polypropylene(PP)
$493.59CAD / Case of 1000
COMPARE
100 5 Polypropylene(PP)
$57.46CAD / Case of 100
COMPARE
100 5 Polypropylene(PP)
$545.15CAD / Case of 1000
COMPARE
150 5 Polypropylene(PP)
$98.72CAD / Case of 100
COMPARE
150 5 Polypropylene(PP)
$1,156.61CAD / Case of 1000
COMPARE
250 10 Polypropylene(PP)
$61.88CAD / Pkg of 50
COMPARE
250 10 Polypropylene(PP)
$729.33CAD / Case of 500
COMPARE
400 25 Polypropylene(PP)
$82.51CAD / Pkg of 50
COMPARE
400 25 Polypropylene(PP)
$950.34CAD / Case of 500
COMPARE
600 25 Polypropylene(PP)
$92.82CAD / Pkg of 25
COMPARE
600 25 Polypropylene(PP)
$1,099.15CAD / Case of 500
Showing 1- 16 of 16